accessibility menu, dialog, popup

UserName

CAS RN 5457-28-3

IUPAC Name: 1H-indole-3-carbonitrile
Synonyms: 3-cyanoindole
Molecular Weight (g/mol): 142.16
Molecular Formula: C9H6N2
InChi Key: CHIFTAQVXHNVRW-UHFFFAOYSA-N
SMILES: N#CC1=CNC2=CC=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)