missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole-3-acetic acid, 99.2%, MP Biomedicals™
$142.25 - $142.25
Chemical Identifiers
| CAS | 87-51-4 |
|---|---|
| Molecular Formula | C10H9NO2 |
| Molecular Weight (g/mol) | 175.19 |
| MDL Number | MFCD00005636 |
| InChI Key | SEOVTRFCIGRIMH-UHFFFAOYSA-N |
| Synonym | indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a |
| PubChem CID | 802 |
| ChEBI | CHEBI:16411 |
| IUPAC Name | 2-(1H-indol-3-yl)acetic acid |
| SMILES | OC(=O)CC1=CNC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN10203725
|
MP Biomedicals Inc
0210203725 |
25 g |
Each for $142.25
|
|
|||||
Chemical Identifiers
| 87-51-4 | |
| 175.19 | |
| SEOVTRFCIGRIMH-UHFFFAOYSA-N | |
| 802 | |
| 2-(1H-indol-3-yl)acetic acid |
| C10H9NO2 | |
| MFCD00005636 | |
| indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a | |
| CHEBI:16411 | |
| OC(=O)CC1=CNC2=CC=CC=C12 |
Specifications
| 87-51-4 | |
| C10H9NO2 | |
| 25 g | |
| SEOVTRFCIGRIMH-UHFFFAOYSA-N | |
| 2-(1H-indol-3-yl)acetic acid | |
| 802 | |
| 175.19 | |
| Powder |
| White | |
| MFCD00005636 | |
| indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a | |
| OC(=O)CC1=CNC2=CC=CC=C12 | |
| 175.19 | |
| CHEBI:16411 | |
| 0.992 | |
| Indole-3-acetic acid, Molecular biology and Cell biology |
Safety and Handling
EINECSNumber : 201-748-2
storageNote1 : Protect from light
Recommended Storage : Store at -0°C