missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-acetic acid, 99.2%, MP Biomedicals™

Catalog No. ICN10203725
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10203725 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10203725 Supplier MP Biomedicals Inc Supplier No. 0210203725
Only null left

Chemical Identifiers

CAS 87-51-4
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00005636
InChI Key SEOVTRFCIGRIMH-UHFFFAOYSA-N
Synonym indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a
PubChem CID 802
ChEBI CHEBI:16411
IUPAC Name 2-(1H-indol-3-yl)acetic acid
SMILES OC(=O)CC1=CNC2=CC=CC=C12

Specifications

Color White
Quantity 25 g
Formula Weight 175.19
Percent Purity 0.992
Physical Form Powder
Chemical Name or Material Indole-3-acetic acid, Molecular biology and Cell biology
EINECSNumber 201-748-2
storageNote1 Protect from light
Recommended Storage Store at -0°C