missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-acetic acid, 98+%

Catalog No. aaa1055622
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1055622 100 g
AAA1055614 25 g
AAA1055636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1055622 Supplier Thermo Scientific Chemicals Supplier No. A1055622
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 87-51-4 | C10H9NO2 | 175.19 g/mol

It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.

Solubility
Soluble in ethanol (50 mg/ml), methanol, DMSO, and chloroform (sparingly). Insoluble in water.

Notes
Light sensitive. Store at -20°C. ncompatible with oxidizing agents.

Chemical Identifiers

CAS 87-51-4
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00005636
InChI Key SEOVTRFCIGRIMH-UHFFFAOYSA-N
Synonym indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a
PubChem CID 802
ChEBI CHEBI:16411
IUPAC Name 2-(1H-indol-3-yl)acetic acid
SMILES OC(=O)CC1=CNC2=CC=CC=C12

Specifications

Melting Point ∼167°C (decomposition)
Flash Point 171°C (339°F)
Quantity 100 g
Beilstein 143358
Sensitivity Light sensitive
Merck Index 14,4964
Solubility Information Soluble in ethanol (50mg/ml),methanol,DMSO,and chloroform (sparingly). Insoluble in water.
Formula Weight 175.19
Percent Purity ≥98%
Chemical Name or Material Indole-3-acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-748-2
RTECSNumber NL3150000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only