missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-acetic Acid, 98%, Spectrum™ Chemical

Catalog No. 18611025
Change view
Click to view available options
Quantity:
5 kg
50 kg
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611025 50 kg Poly Pail
18611024 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611025 Supplier Spectrum Chemical Mfg Cor Supplier No. IN12550KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87-51-4

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 50 kg
Molecular Weight (g/mol) 175.18
Percent Purity 0.98
Grade Ungraded
Physical Form Crystals or Flakes
Chemical Name or Material Indole-3-acetic acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.