missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole-3-acetic Acid, 98%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Poly Pail |
| Quantity | 5 kg |
| Molecular Weight (g/mol) | 175.18 |
| Percent Purity | 0.98 |
| Grade | Ungraded |
| Physical Form | Crystals or Flakes |
| Chemical Name or Material | Indole-3-acetic acid |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.