missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indocyanine green, MP Biomedicals™
Chemical Identifiers
| CAS | 3599-32-4 |
|---|---|
| Molecular Formula | C43H47N2NaO6S2 |
| Molecular Weight (g/mol) | 774.967 |
| MDL Number | MFCD00013078 |
| InChI Key | MOFVSTNWEDAEEK-UHFFFAOYSA-M |
| Synonym | Cardiogreen, Fax green |
| PubChem CID | 132274068 |
| ChEBI | CHEBI:31696 |
| IUPAC Name | sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate |
| SMILES | CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN15502080
|
N/A | 100 mg |
N/A
|
|
|||||
Chemical Identifiers
| 3599-32-4 | |
| 774.967 | |
| MOFVSTNWEDAEEK-UHFFFAOYSA-M | |
| 132274068 | |
| sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate |
| C43H47N2NaO6S2 | |
| MFCD00013078 | |
| Cardiogreen, Fax green | |
| CHEBI:31696 | |
| CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+] |
Specifications
| 3599-32-4 | |
| C43H47N2NaO6S2 | |
| Cardiogreen, Fax green | |
| CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+] | |
| 774.967 | |
| CHEBI:31696 | |
| Green | |
| Indocyanine green |
| ∽ 90% | |
| MFCD00013078 | |
| MOFVSTNWEDAEEK-UHFFFAOYSA-M | |
| sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate | |
| 132274068 | |
| 774.98 | |
| Crystalline Powder |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C). Desiccate.