missing translation for 'onlineSavingsMsg'
Learn More

Indocyanine green, MP Biomedicals™

Catalog No. ICN15502080
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15502080 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15502080 Supplier MP Biomedicals Inc Supplier No. 0215502080

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3599-32-4 | C43H47N2NaO6S2 | 774.967 g/mol

Chemical Identifiers

CAS 3599-32-4
Molecular Formula C43H47N2NaO6S2
Molecular Weight (g/mol) 774.967
MDL Number MFCD00013078
InChI Key MOFVSTNWEDAEEK-UHFFFAOYSA-M
Synonym Cardiogreen, Fax green
PubChem CID 132274068
ChEBI CHEBI:31696
IUPAC Name sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+]

Specifications

Assay Percent Range ∽ 90%
Color Green
pH 6
Quantity 100 mg
Physical Form Crystalline Powder
Formula Weight 774.98
Percent Purity ∼90%
Grade Research
Chemical Name or Material Indocyanine green
Recommended Storage Store at Room Temperature (15 to 30°C). Desiccate.