missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troemner™ Individual Analytical Precision Weights, Class 3
Low magnetic susceptibility, uniform consistency and high resistance to corrosion
Supplier: Troemner™ 80858087
Description
Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.
- Used in extremely stable environments
- 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- 1g to 5kg are two-piece construction with handling knob
- Surfaces are polished to a perfect, mirror like finish
- Appropriate for calibrating balances with moderate precision and with a readability as low as 0.1g to 0.01g
- Weights 1mg through 2kg are supplied in a high-quality polycarbonate case
- Reference standards as well as calibrating Class I balances as specified in OIML R 76
- Calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g
Specifications
| Metric Analytical Weight with Handling Knob | |
| Stainless Steel | |
| 7.95g/cm3 at 20°C | |
| Cylindrical with Handling Knob | |
| Each |
| ASTM Class 3 | |
| 5 kg | |
| 5 kg | |
| Polycarbonate case | |
| 1 |