missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Individual Analytical Precision Weights, Class 3

Low magnetic susceptibility, uniform consistency and high resistance to corrosion

Supplier:  Troemner™ 80858537

 View more versions of this product

Catalog No. 02214032


Only null left
Explore available promotions

Description

Description

Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.

  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • Appropriate for calibrating balances with moderate precision and with a readability as low as 0.1g to 0.01g
  • Weights 1mg through 2kg are supplied in a high-quality polycarbonate case
  • Reference standards as well as calibrating Class I balances as specified in OIML R 76
  • Calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g
Specifications

Specifications

One Piece Metric Analytical Sheet Weight with One Side Turned Up
Stainless Steel
7.95g/cm3 at 20°C
Flat with one end turned up for easy handling
Each
ASTM Class 3
20 mg
20 mg
Polycarbonate case
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions