missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Individual Analytical Precision Weights, Class 3

Low magnetic susceptibility, uniform consistency and high resistance to corrosion

Supplier:  Troemner™ 80858669

 View more versions of this product

Catalog No. 02-214-029


Only null left
Explore available promotions

Description

Description

Designed for your most demanding weighing applications. Analytical Precision Weights meet or exceed specifications as defined in the ANSI/ASTM E617 standard.

  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • Appropriate for calibrating balances with moderate precision and with a readability as low as 0.1g to 0.01g
  • Weights 1mg through 2kg are supplied in a high-quality polycarbonate case
  • Reference standards as well as calibrating Class I balances as specified in OIML R 76
  • Calibrating balances with moderate precision, with a readability as low as 0.1g to 0.01g
Specifications

Specifications

One Piece Metric Analytical Sheet Weight with One Side Turned Up
Aluminum
2.7g/cm3 at 20°C
Flat with one end turned up for easy handling
Each
ASTM Class 3
2 mg
2 mg
Polycarbonate case
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions