missing translation for 'onlineSavingsMsg'
Learn More

Troemner™ Individual Analytical Precision Weights, Class 1

Catalog No. 023191J
Change view
Click to view available options
Mass:
1 mg
2 mg
5 mg
10 mg
50 mg
100 mg
200 mg
300 mg
500 mg
1 g
3 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mass
023191J 200 mg
023191H 500 mg
023191K 100 mg
023191B 50 g
023191P 10 mg
023191E 5 g
023191T 1 mg
023191C 20 g
023191Q 5 mg
023191D 10 g
023191L 50 mg
0221577 200 g
02214021 300 mg
0221595 500 g
023191R 2 mg
023191A 100 g
023191G 1 g
02214022 3 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 023191J

Low magnetic susceptibility, uniform consistency and high resistance to corrosion

  • Used in extremely stable environments
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
  • 1g to 5kg are two-piece construction with handling knob
  • Surfaces are polished to a perfect, mirror like finish
  • Can be used as a reference standard in calibrating other weights
  • Appropriate for calibrating high-precision analytical balances with a readability as low as 0.1 mg to 0.01mg

Certifications

A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements

Specifications

Product Type One Piece Metric Analytical Sheet Weight
Classification ASTM Class 1
Material Stainless Steel
Weight (Metric) 200 mg
Density 7.95g/cm3 at 20°C
Mass 200 mg
Shape Flat with one end turned up for easy handling
Includes Polycarbonate case
Quantity Each
Packaging Quantity 1