missing translation for 'onlineSavingsMsg'
Learn More

Indium wire, 1.0mm (0.04in) dia, Puratronic™, 99.998+% (metals basis)

Catalog No. p-4408115
Change view
Click to view available options
Quantity:
1 m
25 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA11462G1 1 m
AA11462G9 25 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA11462G1 Supplier Thermo Scientific Chemicals Supplier No. 011462G1
Only null left

Form: approx. 5.74g/m | CAS: 7440-74-6 | In | 114.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Silver-White
Melting Point 156.6°C
Boiling Point 2072°C
Appearance Silvery-white wire
Density 7.31g/cm3 at 20°C
Quantity 1 m
Odor Odorless
Form Wire
Assay Percent Range >99.998% (Metals basis)
Chemical Name or Material Indium
EINECSNumber 231-180-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only