missing translation for 'onlineSavingsMsg'
Learn More

Indium wire, 0.5mm (0.02in) dia, Puratronic™, 99.999% (metals basis)

Catalog No. AA001550K
Change view
Click to view available options
Quantity:
0.5 m
2 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA001550K 0.5 m
AA00155G2 2 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA001550K Supplier Thermo Scientific Chemicals Supplier No. 0001550K

CAS: 7440-74-6 | In | 114.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Quantity 0.5 m
Odor Odorless
Weight ≈1.43g/m
Form Wire
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Indium wire, 0.5mm (0.02 in.) dia.
EINECSNumber 231-180-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only