Learn More
Indium wire, 0.25mm (0.01in) dia, Puratronic™, 99.998+% (metals basis)
Form: Approx.0.36g/m | CAS: 7440-74-6 | In | 114.82 g/mol
Supplier: Thermo Scientific Chemicals 040421G6
Description
As solders, thermal fuses, fusible alloys, and in semiconductor industry in InAs and InSb forms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Silver-White | |
| 7440-74-6 | |
| 10 m | |
| >99.998% (Metals basis) | |
| In | |
| indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
| [In] | |
| 114.82 | |
| CHEBI:30430 |
| Indium | |
| Silvery-white wire | |
| Odorless | |
| Wire | |
| MFCD00134048 | |
| APFVFJFRJDLVQX-UHFFFAOYSA-N | |
| indium | |
| 5359967 |
Chemical Identifiers
| 7440-74-6 | |
| 114.82 | |
| APFVFJFRJDLVQX-UHFFFAOYSA-N | |
| 5359967 | |
| indium |
| In | |
| MFCD00134048 | |
| indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
| CHEBI:30430 | |
| [In] |
Safety and Handling
EINECSNumber : 231-180-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only