missing translation for 'onlineSavingsMsg'
Learn More

Indium tin oxide, 99.99% (metals basis)

CAS: 50926-11-9 | InOSn | 249.527 g/mol

$78.38 - $575.49

Chemical Identifiers

CAS 50926-11-9
Molecular Formula InOSn
Molecular Weight (g/mol) 249.527
MDL Number MFCD00171662
InChI Key AMGQUBHHOARCQH-UHFFFAOYSA-N
Synonym acmc-1au9m, tin ii oxide indigane
PubChem CID 57350325
IUPAC Name indium;oxotin
SMILES O=[Sn].[In]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3634806
View Documents
Thermo Scientific Chemicals
03634806
5 g
Each for $78.38
Only null left
 
AA3634814
View Documents
Thermo Scientific Chemicals
03634814
25 g
Each for $204.78
Only null left
 
AA3634822
View Documents
Thermo Scientific Chemicals
03634822
100 g
Each for $575.49
Only null left
 
Description

Description

Indium tin oxide is used in nanotechnology to provide a path to a new generation of solar cells, to make transparent conductive coatings for displays such as liquid crystal displays, flat panel displays, plasma displays, touch panels, and electronic ink. Thin films of ITO are also used in organic light-emitting diodes, antistatic coatings and EMI shielding. In organic light-emitting diodes, ITO is used as the anode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50926-11-9
249.527
AMGQUBHHOARCQH-UHFFFAOYSA-N
57350325
O=[Sn].[In]
InOSn
MFCD00171662
acmc-1au9m, tin ii oxide indigane
indium;oxotin

Specifications

White
Powder
99.99% (Metal basis)
MFCD00171662
Insoluble in water.
O=[Sn].[In]
249.527
Odorless
Indium tin oxide
50926-11-9
5 g
InOSn
acmc-1au9m, tin ii oxide indigane
AMGQUBHHOARCQH-UHFFFAOYSA-N
indium;oxotin
57350325
White powder
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P271-P281-P304+P340-P308+P313-P501c

H335-H351

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only