CAS RN 50926-11-9
IUPAC Name:
stannanone indium
Synonyms:
tin(II) oxide indium
Molecular Weight (g/mol):
249.53
Molecular Formula:
InOSn
InChi Key:
AMGQUBHHOARCQH-UHFFFAOYSA-N
SMILES:
[In].O=[Sn]
1
–
2
of
2
results
1
Indium tin oxide, 99.99% (metals basis)
CAS: 50926-11-9 Molecular Formula: InOSn Molecular Weight (g/mol): 249.527 MDL Number: MFCD00171662 InChI Key: AMGQUBHHOARCQH-UHFFFAOYSA-N Synonym: acmc-1au9m,tin ii oxide indigane PubChem CID: 57350325 IUPAC Name: indium;oxotin SMILES: O=[Sn].[In]
2
Indium tin oxide, NanoTek∣r, 99.5%
CAS: 50926-11-9 Molecular Formula: InOSn Molecular Weight (g/mol): 249.527 MDL Number: MFCD00171662 InChI Key: AMGQUBHHOARCQH-UHFFFAOYSA-N Synonym: acmc-1au9m,tin ii oxide indigane PubChem CID: 57350325 IUPAC Name: indium;oxotin SMILES: O=[Sn].[In]