Learn More
Indium tin oxide, 99.99% (metals basis)
Description
Indium tin oxide is used in nanotechnology to provide a path to a new generation of solar cells, to make transparent conductive coatings for displays such as liquid crystal displays, flat panel displays, plasma displays, touch panels, and electronic ink. Thin films of ITO are also used in organic light-emitting diodes, antistatic coatings and EMI shielding. In organic light-emitting diodes, ITO is used as the anode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99.99% (Metal basis) |
| Solubility Information | Insoluble in water. |
| Odor | Odorless |
| Appearance | White powder |
| Chemical Name or Material | Indium tin oxide |
Safety and Handling
| Hazard Category | H335-H351 |
| Precautionary Statement | P201-P202-P261-P271-P281-P304+P340-P308+P313-P501c |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only