missing translation for 'onlineSavingsMsg'
Learn More

Indium powder, -325 mesh, Puratronic™, 99.995% (metals basis)

Catalog No. AA0015114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0015114 25 g
AA0015106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA0015114 Supplier Thermo Scientific Chemicals Supplier No. 00015114

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-74-6 | In | 114.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Silver to Gray
Melting Point 156.6°C
Boiling Point 2072°C
Appearance Silver-gray powder
Density 7.31
Quantity 25 g
Mesh Size -325 Mesh
Odor Odorless
Form Powder
Assay Percent Range 99.999% (Metals basis)
Formula Weight 114.82
Chemical Name or Material Indium
Show More Show Less
Hazard Category H228-H302+H312+H332
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.1; Packaging Group: II
EINECSNumber 231-180-0
RTECSNumber NL1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only