missing translation for 'onlineSavingsMsg'
Learn More

Indium powder, -325 mesh, 99.99% (metals basis)

Catalog No. p-7048612
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
AA1102406 5 g
AA1102414 25 g
2 options

Catalog No. AA1102406

Supplier: Thermo Scientific Chemicals 01102406

Only null left

CAS: 7440-74-6 | In | 114.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Silver to Gray
Melting Point 156.6°C
Boiling Point 2072°C
Density 7.31 g/mL
Quantity 5 g
Mesh Size −325 Mesh
Odor Odorless
Form Powder
UN Number UN3089
Formula Weight 114.82
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Indium powder
Show More Show Less
Hazard Category H228-H302+H312+H332
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 231-180-0
RTECSNumber NL1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only