missing translation for 'onlineSavingsMsg'
Learn More

Indium ingot, polycrystalline, Puratronic™, 99.9995% (metals basis)

Catalog No. p-7048463
Change view
Click to view available options
Quantity:
50 g
5 x 50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1061618 50 g
AA1061690 5 x 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1061618 Supplier Thermo Scientific Chemicals Supplier No. 1061618
Only null left

CAS: 7440-74-6 | In | 114.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Melting Point 156.6°C
Boiling Point 2072°C
Density 7.310 g/mL
Quantity 50 g
Odor Odorless
Form Ingot
Formula Weight 114.818
Percent Purity 99.9995%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Indium ingot
Show More Show Less
EINECSNumber 231-180-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only