missing translation for 'onlineSavingsMsg'
Learn More

Indium ingot/button,≈31mm (1.2 in.) dia. x 5.3mm (0.21 in.) thick, 99.99% (metals basis), Thermo Scientific™

Catalog No. AA45111KF
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45111KF 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45111KF Supplier Thermo Scientific Chemicals Supplier No. 45111KF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Silver
Quantity 1 Pc.
Odor Odorless
Weight ≈28g/piece
Form Pieces
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Indium ingot/button, ≈31mm (1.2 in.) dia. x 5.3mm (0.21 in.) thick
EINECSNumber 231-180-0
RTECSNumber NL1050000
TSCA Yes
Recommended Storage Ambient temperatures