Learn More
Indium(III) sulfide, 99.995% (metals basis)
CAS: 12030-24-9 | In2S3 | 325.82 g/mol
$1412.25 - $1412.25
Chemical Identifiers
| CAS | 12030-24-9 |
|---|---|
| Molecular Formula | In2S3 |
| Molecular Weight (g/mol) | 325.82 |
| MDL Number | MFCD00011061 |
| InChI Key | SIXIBASSFIFHDK-UHFFFAOYSA-N |
| IUPAC Name | diindium(3+) trisulfanediide |
| SMILES | [S--].[S--].[S--].[In+3].[In+3] |
Description
Indium(III) sulfide emerged as a promising low-hazard buffer material and is used to improve the properties of solar cells and also replaces the hazardous CdS. It is a mid band gap semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12030-24-9 | |
| 325.82 | |
| SIXIBASSFIFHDK-UHFFFAOYSA-N | |
| [S--].[S--].[S--].[In+3].[In+3] |
| In2S3 | |
| MFCD00011061 | |
| diindium(3+) trisulfanediide |
Specifications
| 1050°C | |
| 12030-24-9 | |
| 50 g | |
| MFCD00011061 | |
| SIXIBASSFIFHDK-UHFFFAOYSA-N | |
| diindium(3+) trisulfanediide | |
| 325.83 | |
| Like rotten eggs | |
| Indium (III) sulfide |
| Red to Orange | |
| Powder | |
| In2S3 | |
| Insoluble in water and organic solvents. | |
| [S--].[S--].[S--].[In+3].[In+3] | |
| 325.82 | |
| 99.995% | |
| Moisture sensitive | |
| (metals basis) |
Safety and Handling
GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
EINECSNumber : 234-742-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only