Learn More
Indium(III) sulfide, 99.98% (metals basis)
CAS: 12030-24-9 | In2S3 | 325.82 g/mol
$78.56 - $1286.90
Chemical Identifiers
| CAS | 12030-24-9 |
|---|---|
| Molecular Formula | In2S3 |
| Molecular Weight (g/mol) | 325.82 |
| MDL Number | MFCD00011061 |
| InChI Key | SIXIBASSFIFHDK-UHFFFAOYSA-N |
| IUPAC Name | diindium(3+) trisulfanediide |
| SMILES | [S--].[S--].[S--].[In+3].[In+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4483504
|
Thermo Scientific Chemicals
04483504 |
2 g |
Each for $78.56
|
|
|||||
|
AA4483509
|
Thermo Scientific Chemicals
04483509 |
10 g |
Each for $280.68
|
|
|||||
|
AA4483518
|
Thermo Scientific Chemicals
04483518 |
50 g |
Each for $1,286.90
|
|
|||||
Description
Indium(III) sulfide acts as mid band gap semiconductor. It is an important material for optoelectronic and photovoltaic applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12030-24-9 | |
| 325.82 | |
| SIXIBASSFIFHDK-UHFFFAOYSA-N | |
| [S--].[S--].[S--].[In+3].[In+3] |
| In2S3 | |
| MFCD00011061 | |
| diindium(3+) trisulfanediide |
Specifications
| 1050°C | |
| Powder | |
| In2S3 | |
| Insoluble in water. | |
| [S--].[S--].[S--].[In+3].[In+3] | |
| 325.82 | |
| 99.98% | |
| Moisture sensitive | |
| (metals basis) |
| 12030-24-9 | |
| 2 g | |
| MFCD00011061 | |
| SIXIBASSFIFHDK-UHFFFAOYSA-N | |
| diindium(3+) trisulfanediide | |
| 325.83 | |
| Like rotten eggs | |
| Indium (III) sulfide |
Safety and Handling
GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
EINECSNumber : 234-742-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only