Learn More
Indium(III) phosphide, polycrystalline lump, 99.99% (metals basis)
CAS: 22398-80-7 | InP | 145.792 g/mol
$176.53 - $630.22
Chemical Identifiers
| CAS | 22398-80-7 |
|---|---|
| Molecular Formula | InP |
| Molecular Weight (g/mol) | 145.792 |
| MDL Number | MFCD00016153 |
| InChI Key | GPXJNWSHGFTCBW-UHFFFAOYSA-N |
| Synonym | indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875 |
| PubChem CID | 31170 |
| IUPAC Name | indiganylidynephosphane |
| SMILES | P#[In] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1550903
|
Thermo Scientific Chemicals
A1550903 |
1 g |
Each for $176.53
|
|
|||||
|
AAA1550906
|
Thermo Scientific Chemicals
A1550906 |
5 g |
Each for $630.22
|
|
|||||
Description
InP is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct bandgap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsInP is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct bandgap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.
Solubility
Insoluble in water.
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Chemical Identifiers
| 22398-80-7 | |
| 145.792 | |
| GPXJNWSHGFTCBW-UHFFFAOYSA-N | |
| 31170 | |
| P#[In] |
| InP | |
| MFCD00016153 | |
| indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875 | |
| indiganylidynephosphane |
Specifications
| 1070°C | |
| Polycrystalline Lump | |
| InP | |
| 14,4953 | |
| Insoluble in water. | |
| P#[In] | |
| 145.792 | |
| 145.79 | |
| 4.81 | |
| (metals basis) |
| 22398-80-7 | |
| 1 g | |
| MFCD00016153 | |
| indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875 | |
| GPXJNWSHGFTCBW-UHFFFAOYSA-N | |
| indiganylidynephosphane | |
| 31170 | |
| 99.99% | |
| Indium(III) phosphide |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P260-P264b-P270-P281-P308+P313-P501c
H350-H361f-H372
EINECSNumber : 244-959-5
RTECSNumber : NL1800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only