accessibility menu, dialog, popup

UserName

PIn

CAS RN 22398-80-7

IUPAC Name: indiganylidynephosphane
Synonyms: indium phosphide
Molecular Weight (g/mol): 145.79
Molecular Formula: InP
InChi Key: GPXJNWSHGFTCBW-UHFFFAOYSA-N
SMILES: P#[In]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)