missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) phosphide, polycrystalline lump, 99.99% (metals basis)

Catalog No. AAA1550903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1550903 1 g
AAA1550906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1550903 Supplier Thermo Scientific Chemicals Supplier No. A1550903
Only null left

CAS: 22398-80-7 | InP | 145.792 g/mol

InP is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct bandgap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
InP is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct bandgap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.

Chemical Identifiers

CAS 22398-80-7
Molecular Formula InP
Molecular Weight (g/mol) 145.792
MDL Number MFCD00016153
InChI Key GPXJNWSHGFTCBW-UHFFFAOYSA-N
Synonym indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875
PubChem CID 31170
IUPAC Name indiganylidynephosphane
SMILES P#[In]

Specifications

Melting Point 1070°C
Physical Form Polycrystalline Lump
Quantity 1 g
Merck Index 14,4953
Solubility Information Insoluble in water.
Formula Weight 145.79
Percent Purity 99.99%
Density 4.81
Assay (metals basis)
Chemical Name or Material Indium(III) phosphide
Hazard Category H350-H361f-H372
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P308+P313-P501c
EINECSNumber 244-959-5
RTECSNumber NL1800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only