missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium(III) phosphide, 99.999% (metals basis)
Description
The water soluble InP/ZnS core/shell QDs are a safer alternative to CdSe/ZnS QDs for biological applications, by comparing their toxicity in vitro (cell culture) and in vivo.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1070°C |
| Quantity | 5 g |
| Merck Index | 14,4953 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 145.79 |
| Percent Purity | 99.999% |
| Density | 4.81 g/mL |
| Assay | (metals basis) |
| Chemical Name or Material | Indium(III) phosphide |
Safety and Handling
| Hazard Category | H350-H361f-H372 |
| Hazard Statement | GHS H Statement H351 Suspected of causing cancer. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P281-P308+P313-P501c |
| EINECSNumber | 244-959-5 |
| RTECSNumber | NL1800000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only