missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) oxide, 99.994% (metals basis)

CAS: 1312-43-2 | In2O3 | 277.63 g/mol

Supplier:  Thermo Scientific Chemicals 01185309

 View more versions of this product

Catalog No. AA1185309


Only null left
Explore available promotions

Description

Description

Indium(III) oxide is used as a chemical reagent and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Indium(III) oxide
Yellow
Powder
99.994% (Metals basis)
MFCD00011060
PJXISJQVUVHSOJ-UHFFFAOYSA-N
diindium(3+) trioxidandiide
277.64
7.18
850° volat.
1312-43-2
10 g
In2O3
Insoluble in water.
[O--].[O--].[O--].[In+3].[In+3]
277.63
Odorless

Chemical Identifiers

1312-43-2
277.63
PJXISJQVUVHSOJ-UHFFFAOYSA-N
[O--].[O--].[O--].[In+3].[In+3]
In2O3
MFCD00011060
diindium(3+) trioxidandiide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-193-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only