missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) oxide, 99.994% (metals basis)

Catalog No. p-4781941
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
250 g
50 g
Catalog No. Quantity
AA1185309 10 g
AA1185318 50 g
AA1185330 250 g
3 options

Catalog No. AA1185309

Supplier: Thermo Scientific Chemicals 01185309

Only null left

CAS: 1312-43-2 | In2O3 | 277.63 g/mol

Indium(III) oxide is used as a chemical reagent and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Melting Point 850° volat.
Color Yellow
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.994% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 277.64
Odor Odorless
Density 7.18
Chemical Name or Material Indium(III) oxide
EINECSNumber 215-193-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only