missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) Chloride Anhydrous 99.0+%, TCI America™

Catalog No. p-7131593
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07785G 5 g
I077825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07785G Supplier TCI America Supplier No. I07785G
Only null left

Chemical Identifiers

CAS 10025-82-8
Molecular Formula Cl3In
MDL Number MFCD00011058

Specifications

Melting Point 586°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
UN Number 3260
Formula Weight 221.17
Percent Purity ≥99.0% (T)
Chemical Name or Material Indium(III) Chloride Anhydrous
EINECSNumber (1)-0227
RTECSNumber NL1400000
TSCA Yes