missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium(III) chloride, anhydrous, 99.999% (metals basis)
Description
It is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 800°C |
| Melting Point | 586°C |
| Color | White |
| Physical Form | Crystalline |
| Quantity | 1 g |
| UN Number | UN3260 |
| Merck Index | 14,4958 |
| Solubility Information | It is very soluble in water and THF. It is sparingly soluble in alcohol and insoluble in ether. |
| Formula Weight | 221.18 |
| Percent Purity | 99.999% |
| Show More |
Safety and Handling
| Hazard Category | H302-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S. |
| EINECSNumber | 233-043-0 |
| RTECSNumber | NL1400000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only