Learn More
Indium(III) chloride, anhydrous, 99.99% (metals basis)
CAS: 10025-82-8 | InCl3
Supplier: Thermo Scientific Chemicals 04197730
Description
Indium(III) chloride is used in organic reactions such as Friedel-crafts acylation and Diels-Alder reaction as a Lewis acid catalyst. It is an efficient catalyst for thioacetalization and its chemoselectivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 800°C | |
| 586°C | |
| 10025-82-8 | |
| 250 g | |
| InCl3 | |
| Soluble in water and tetrhydrofuran. Sparingly soluble in alcohol. Insoluble in ether. | |
| Hygroscopic |
| Indium(III) chloride | |
| White | |
| Crystals | |
| 99.99% (Metals basis) | |
| MFCD00011058 | |
| 221.18 | |
| 3.46 |
Chemical Identifiers
| 10025-82-8 | |
| InCl3 | |
| MFCD00011058 |
Safety and Handling
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 233-043-0
RTECSNumber : NL1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only