Learn More
Indium(III) bromide, 99.99%, (trace metal basis)
CAS: 13465-09-3 | Br3In | 354.52 g/mol
$304.46 - $304.46
Chemical Identifiers
| CAS | 13465-09-3 |
|---|---|
| Molecular Formula | Br3In |
| Molecular Weight (g/mol) | 354.52 |
| MDL Number | MFCD00011057 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC389030100
|
Thermo Scientific Chemicals
389030100 |
10 g | Glass bottle |
Each for $304.46
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisChemical Identifiers
| 13465-09-3 | |
| Br3In | |
| 354.52 | |
| MFCD00011057 |
Specifications
| 656.0°C | |
| White | |
| Crystalline Powder | |
| 99.99% | |
| InBr3 | |
| 15, 4997 | |
| 354.52 | |
| Trace Metal Basis | |
| Indium(III) bromide |
| 220.0°C | |
| 13465-09-3 | |
| 10 g | |
| Br3In | |
| MFCD00011057 | |
| 354.52 | |
| 99.99% | |
| Glass bottle |
Safety and Handling
GHS H Statement
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
EINECSNumber : 236-692-8
RUO – Research Use Only