accessibility menu, dialog, popup

UserName

CAS RN 13465-09-3

BrBrBrIn

CAS RN 13465-09-3

IUPAC Name: indium(3+) tribromide
Synonyms: indium(III) bromide
Molecular Weight (g/mol): 354.53
Molecular Formula: Br3In
InChi Key: JKNHZOAONLKYQL-UHFFFAOYSA-K
SMILES: [Br-].[Br-].[Br-].[In+3]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)