missing translation for 'onlineSavingsMsg'
Learn More

Indium foil, 2.0mm (0.08in) thick, Puratronic™, 99.998% (metals basis)

CAS: 7440-74-6 | In | 114.82 g/mol

$421.90 - $739.66

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA11382FF
View Documents
Thermo Scientific Chemicals
011382FF
25 x 25 mm
Each for $421.90
Only null left
 
AA11382FI
View Documents
Thermo Scientific Chemicals
011382FI
50 x 50 mm
Each for $739.66
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-74-6
114.82
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
CHEBI:30430
[In]

Specifications

7440-74-6
Odorless
Foil
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
5359967
99.998%
Indium foil, 2.0mm (0.08 in.) thick
25 x 25 mm
≈9.1g/25x25mm
In
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
[In]
114.82
CHEBI:30430
Puratronic
(metals basis)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-180-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only