missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium AA Standard, 1000 ppm In in 3% HNO3, Ricca Chemical
Chemical Identifiers
| CAS | 7440-74-6 |
|---|---|
| Molecular Formula | In |
| Molecular Weight (g/mol) | 114.82 |
| MDL Number | MFCD00134048 |
| InChI Key | APFVFJFRJDLVQX-UHFFFAOYSA-N |
| IUPAC Name | indium |
| SMILES | [In] |
| Catalog Number | Mfr. No. | Packaging | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Packaging | Price | Quantity | |||||
|
AIN1KN-100
|
Ricca Chemical Company
AIN1KN100 |
Natural LDPE Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AIN1KN-500
|
Ricca Chemical Company
AIN1KN500 |
Natural Poly Bottle |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Specifications
| Natural LDPE Bottle | |
| 90.0,8.1,0.09 | |
| 93.0,8.43,0.09 | |
| MFCD00134048 | |
| [In] | |
| 114.82 |
| 7440-74-6,7697-37-2,7732-18-5 | |
| 1,000ppm | |
| In | |
| APFVFJFRJDLVQX-UHFFFAOYSA-N | |
| indium | |
| Indium AA Standard |