missing translation for 'onlineSavingsMsg'
Learn More

Indium, 99.99%, (trace metal basis), foil, 0.127 mm thick

Catalog No. p-3776743
Change view
Click to view available options
Quantity:
2.3 g
9.2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC318110023 2.3 g
AC318110092 9.2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC318110023 Supplier Thermo Scientific Chemicals Supplier No. 318110023
Only null left

CAS: 7440-74-6 | In | 114.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.82
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]

Specifications

Color Gray
Melting Point 156.0°C
Boiling Point 2000.0°C
Quantity 2.3 g
Form Foil
Assay Percent Range 99.99%
Linear Formula In
Fieser 06,293; 14,181; 16,181; 17,152
Merck Index 15, 4987
Formula Weight 114.82
Percent Purity 99.99%
Grade Trace Metal Basis
Chemical Name or Material Indium
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only