Learn More
Thermo Scientific Chemicals Indigo Carmine, pure, certified
CAS: 860-22-0 | C16H8N2Na2O8S2 | 466.35 g/mol
$102.85 - $288.37
Chemical Identifiers
| CAS | 860-22-0 |
|---|---|
| Molecular Formula | C16H8N2Na2O8S2 |
| Molecular Weight (g/mol) | 466.35 |
| MDL Number | MFCD00005723 |
| InChI Key | KHLVKKOJDHCJMG-QDBORUFSSA-L |
| Synonym | Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt |
| PubChem CID | 5284351 |
| IUPAC Name | disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate |
| SMILES | [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC412300250
|
Thermo Scientific Chemicals
41230 0250 |
25 g | Glass bottle |
Each for $102.85
|
|
||||
|
AC412301000
|
Thermo Scientific Chemicals
412301000 |
100 g | Glass bottle |
Each for $288.37
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 860-22-0 | |
| 466.35 | |
| KHLVKKOJDHCJMG-QDBORUFSSA-L | |
| 5284351 | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
| C16H8N2Na2O8S2 | |
| MFCD00005723 | |
| Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt | |
| disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate |
Specifications
| 860-22-0 | |
| 80% min. | |
| MFCD00005723 | |
| 15, 4982 | |
| Solubility in water: 1g/100mL (25 c). Other solubilities: slightly soluble in alcohol, practically insoluble in most other organic, solvents | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O | |
| disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate | |
| 5284351 | |
| 466.34 | |
| Blue to Purple | |
| Indigo carmine, Pure |
| 0.98 to 1.07 | |
| C16H8N2Na2O8S2 | |
| 25, 304 | |
| Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt | |
| KHLVKKOJDHCJMG-QDBORUFSSA-L | |
| Authentic | |
| 466.35 | |
| 608 to 612nm | |
| Pure | |
| Fine Crystalline Powder |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only