missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Indigo Carmine, pure, certified
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Absorption Ratio | 0.98 to 1.07 |
| Dye Content | 80% min. |
| Infrared Spectrum | Authentic |
| Lambda Max | 608 to 612nm |
| Packaging | Glass bottle |
| Beilstein | 25, 304 |
| Merck Index | 15, 4982 |
| Color | Blue to Purple |
| Solubility Information | Solubility in water: 1g/100mL (25 c). Other solubilities: slightly soluble in alcohol, practically insoluble in most other organic, solvents |
| Quantity | 25 g |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement: Harmful if swallowed. |
| Precautionary Statement | GHS P Statement: IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only