missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indigo Carmine, pure, certified

CAS: 860-22-0 | C16H8N2Na2O8S2 | 466.35 g/mol

Supplier:  Thermo Scientific Chemicals 412301000

 View more versions of this product

Catalog No. AC412301000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Indigo carmine
860-22-0
80% min.
MFCD00005723
15, 4982
Solubility in water: 1g/100mL (25 c). Other solubilities: slightly soluble in alcohol, practically insoluble in most other organic, solvents
Authentic
disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate
5284351
466.34
Glass bottle
100 g
Pure
0.98 to 1.07
C16H8N2Na2O8S2
25, 304
Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt
KHLVKKOJDHCJMG-QDBORUFSSA-L
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
466.35
608 to 612nm
Pure
Blue to Purple
Fine Crystalline Powder

Chemical Identifiers

860-22-0
466.35
KHLVKKOJDHCJMG-QDBORUFSSA-L
5284351
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
C16H8N2Na2O8S2
MFCD00005723
Acid Blue 74, C.I. 73015, 5, 5'-Indigodisulfonic acid, disodium salt
disodium;(2E)-3-oxo-2-(3-oxo-5-sulfonato-1H-indol-2-ylidene)-1H-indole-5-sulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only