missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indeno[1,2,3-cd]pyrene, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| CAS | 193-39-5 |
| Physical Form | Neat |
| Linear Formula | C22H12 |
| MDL Number | MFCD00152577 |
| Type | Organic Pollutant Standard |
| Quantity | 10 mg |
| Formula Weight | 276.33 |
| Grade | Analytical Standard |