missing translation for 'onlineSavingsMsg'
Learn More

Indene, 90%, technical, stabilized

Catalog No. p-148191
Change view
Click to view available options
Quantity:
5 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC122140050 5 g Glass bottle
AC122142500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC122140050 Supplier Thermo Scientific Chemicals Supplier No. 122140050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-13-6 | C9H8 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-13-6 , 98-29-3
Molecular Formula C9H8
Molecular Weight (g/mol) 116.16
MDL Number MFCD00003777
InChI Key YBYIRNPNPLQARY-UHFFFAOYSA-N
Synonym indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy
PubChem CID 7219
ChEBI CHEBI:41921
IUPAC Name 1H-indene
SMILES C1C=CC2=CC=CC=C21

Specifications

Melting Point -5°C to 3°C
Color White to Yellow
Density 0.9900g/mL
Boiling Point 181.0°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5670 to 1.5720
Quantity 5 g
Beilstein 05, 515
Specific Gravity 0.99
Merck Index 15, 4978
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with most organic solvents,soluble in acetone,miscible with ethanol and ethyl ether
Stabilizer 5 to 10ppm tert-butylcatechol
Formula Weight 116.16
Percent Purity 90%
Grade Technical
Physical Form Crystalline Powder
Chemical Name or Material Indene, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 202-393-6
RTECSNumber NK8225000
TSCA TSCA

RUO – Research Use Only