missing translation for 'onlineSavingsMsg'
Learn More

Indazole 99.0+%, TCI America™

Catalog No. I02781G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02781G 1 g
I02785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I02781G Supplier TCI America Supplier No. I02781G

Chemical Identifiers

CAS 271-44-3
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00005691
InChI Key BAXOFTOLAUCFNW-UHFFFAOYSA-N
Synonym indazole, 1h-benzopyrazole, 2-azaindole, isoindazole, 2h-indazole, 1,2-diazaindene, 1,2-benzodiazole, 1h-indazol, benzopyrazole, 1,2-benzopyrazole
PubChem CID 9221
ChEBI CHEBI:36669
IUPAC Name 1H-indazole
SMILES C1=CC=C2C(=C1)C=NN2

Specifications

Melting Point 147°C
Color White
Boiling Point 270°C
Quantity 1 g
Formula Weight 118.14
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Indazole
RTECSNumber NK7745000
TSCA No