accessibility menu, dialog, popup

UserName

CAS RN 271-44-3

IUPAC Name: 1H-indazole
Synonyms: indazole isoindazole 1H-benzopyrazole
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: BAXOFTOLAUCFNW-UHFFFAOYSA-N
SMILES: N1N=CC2=CC=CC=C12
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)