missing translation for 'onlineSavingsMsg'
Learn More

Indane, 95%

Catalog No. AC292481000
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC292481000 100 g Glass bottle
AC292480050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC292481000 Supplier Thermo Scientific Chemicals Supplier No. 292481000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 496-11-7 | C9H10 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 496-11-7
MDL Number MFCD00003795
InChI Key PQNFLJBBNBOBRQ-UHFFFAOYSA-N
Synonym indan, indane, hydrindene, benzocyclopentane, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, 1,2-hydrindene, hydrindonaphthene, indane alkane, indene, 2,3-dihydro
PubChem CID 10326
ChEBI CHEBI:37911
IUPAC Name 2,3-dihydro-1H-indene
SMILES C1CC2=CC=CC=C2C1

Specifications

Melting Point -51°C
Color Colorless to Yellow
Density 0.9600g/mL
Boiling Point 176°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C9H10
Refractive Index 1.5350 to 1.538
Quantity 100 g
Beilstein 05,II,376
Specific Gravity 0.96
Merck Index 15, 4971
Solubility Information Solubility in water: insoluble. Other solubilities: misciple in ethanol,diethyl ether;,slightly soluble in chloroform
Molecular Weight (g/mol) 118.18
Formula Weight 118.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Indane, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-814-7
RTECSNumber NK3750000
TSCA TSCA

RUO – Research Use Only