missing translation for 'onlineSavingsMsg'
Learn More

Indane, 95%

Catalog No. AAB21442AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21442AP 500 mL
AAB21442AE 100 mL
AAB214420F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB21442AP Supplier Thermo Scientific Chemicals Supplier No. B21442AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 496-11-7 | C9H10 | 118.179 g/mol

Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.

Solubility
Soluble in ethanol, diethyl ether, chloroform (slightly), and ether. Insoluble in water.

Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 496-11-7
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00003795
InChI Key PQNFLJBBNBOBRQ-UHFFFAOYSA-N
Synonym indan, indane, hydrindene, benzocyclopentane, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, 1,2-hydrindene, hydrindonaphthene, indane alkane, indene, 2,3-dihydro
PubChem CID 10326
ChEBI CHEBI:37911
IUPAC Name 2,3-dihydro-1H-indene
SMILES C1CC2=CC=CC=C2C1

Specifications

Melting Point -51°C
Density 0.962
Boiling Point 175°C to 177°C
Flash Point 50°C (122°F)
Refractive Index 1.537
Quantity 500 mL
UN Number UN3295
Beilstein 1904376
Merck Index 14,4931
Solubility Information Soluble in ethanol,diethyl ether,chloroform (slightly),and ether. Insoluble in water.
Formula Weight 118.18
Percent Purity 95%
Chemical Name or Material Indane
Show More Show Less
Hazard Category H226-H304
Hazard Statement GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 207-814-7
RTECSNumber NK3750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only