missing translation for 'onlineSavingsMsg'
Learn More

1-Acetylpyrene, 97%

Catalog No. AC449300050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449300050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC449300050 Supplier Thermo Scientific Chemicals Supplier No. 449300050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3264-21-9 | C18H12O | 244.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3264-21-9
Molecular Formula C18H12O
Molecular Weight (g/mol) 244.29
InChI Key KCIJNJVCFPSUBQ-UHFFFAOYSA-N
Synonym 1-acetylpyrene, ethanone, 1-1-pyrenyl, 1-pyren-1-yl ethanone, ethanone,1-1-pyrenyl, acetylpyrene, 1-1-pyrenyl ethanone #, acmc-1cs50
PubChem CID 96251
IUPAC Name 1-pyren-1-ylethanone
SMILES CC(=O)C1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1

Specifications

Melting Point 86.0°C to 89.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 244.29
Percent Purity 97%
Chemical Name or Material 1-Acetylpyrene

RUO – Research Use Only