missing translation for 'onlineSavingsMsg'
Learn More

Indane-5-sulfonyl chloride, 97%

Catalog No. p-7036204
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54462MD 250 mg
AAH5446203 1 g
AAH5446206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH54462MD Supplier Thermo Scientific Chemicals Supplier No. H54462MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52205-85-3 | C9H9ClO2S | 216.679 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52205-85-3
Molecular Formula C9H9ClO2S
Molecular Weight (g/mol) 216.679
MDL Number MFCD05237217
InChI Key SWLIXMXSCZYVTQ-UHFFFAOYSA-N
Synonym indane-5-sulfonyl chloride, indan-5-sulfonyl chloride, indan-5-sulfonylchloride, 5-indanesulfonoyl chloride, chloroindan-5-ylsulfone, 1h-indene-5-sulfonyl chloride, 2,3-dihydro, indanesulfonoylchloride, pubchem5538, 5-chlorosulphonyl indane, acmc-1av5j
PubChem CID 3142583
IUPAC Name 2,3-dihydro-1H-indene-5-sulfonyl chloride
SMILES C1CC2=C(C1)C=C(C=C2)S(=O)(=O)Cl

Specifications

Quantity 250 mg
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 216.69
Percent Purity 97%
Chemical Name or Material Indane-5-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only