missing translation for 'onlineSavingsMsg'
Learn More

Indan Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022523
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022523 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022523 Supplier Merck Emd Millipore Supplier No. 45789100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 496-11-7
Melting Point -51°C (literature)
Density 0.965 g/mL (at 25°C (literature))
Boiling Point 176°C (literature)
Flash Point 54°C (Closed Cup)
Molecular Formula C9H10
Refractive Index n20/D 1.537 (literature)
Linear Formula C9H10
MDL Number MFCD00003795
Quantity 100 mg
UN Number UN 3295 C 3 / PGIII
Synonym Hydrindene
Formula Weight 118.18
Grade Analytical Standard
Physical Form Neat
Show More Show Less